All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H24O2
CAS Number:
Molecular Weight:
200.32
FEMA Number:
4749
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens
Recommended Products
biological source
synthetic
Quality Level
grade
Halal
reg. compliance
FDA 21 CFR 117
Assay
≥95%
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
creamy; tropical; fruity; sweet
SMILES string
O=C(CCC(C)C)OCCCC(C)C
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
141.8 °F
Flash Point(C)
61 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service