Skip to Content
Merck
All Photos(1)

Key Documents

NIST956E

Electrolytes in Frozen Human Serum

new

NIST®SRM® 956e

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

liquid

packaging

pkg of 6 × 2 mL

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

General description

A unit of SRM 956e consists of six sealed borosilicate glass ampoules of frozen human serum, two ampoules each of three different concentration levels: Level 1, Level 2, and Level 3. Each ampoule contains approximately 2 mL of frozen human serum. The certified values delivered by this Standard Reference Material (SRM) are intended for the calibration and validation of procedures and methods employed in clinical analysis for the determination of electrolytes in either diluted or undiluted human serum or plasma. SRM 956e_cert SRM 956e _SDS

Application

The electrolytes in frozen human serum are primarily intended for use in the calibration and validation of procedures and methods employed in clinical analysis for the determination of electrolytes in either diluted or undiluted human serum or plasma. This SRM can be used for calibrating direct-reading ion-selective electrode (ISE) analyzers and for quality assurance in validating secondary reference materials.

Other Notes

  • Notes on Biomaterials, disposal, and transport are available on NIST MSDS.
  • Information on expiry, storage, and usage is provided in the NIST certificate.
Certified for the analytes listed below. Calcium (Ca), Chloride (Cl), Lithium (Li), Magnesium (Mg), Potassium (K), Sodium (Na)

Reference/Informational value is provided for the following analytes. Ionized Calcium (Ca2+, 12 H2O), Phosphate (PO43-), Phosphorus (P)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service