Skip to Content
Merck
All Photos(1)

Key Documents

CCW4693

Millipore

YSG Medium in flasks

Solid medium for the detection and isolation of Alicyclobacillus, in fruit juices and other acidic food, suitable for microbiology

Synonym(s):

YSGM, Yeast Starch Glucose Agar, Yeast-Starch-Glucose medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
Pricing and availability is not currently available.

Quality Level

form

solid

shelf life

18 mo. at 2‑14 °C (limited shelf life, expiry date on the label)

feature

ready-to-use

composition

agar, 15.0 g/L
glucose, 1.0 g/L
soluble starch, 2.0 g/L
yeast extract, 2.0 g/L

packaging

box of 10 bottles

storage condition

(Tightly closed)

parameter

200 mL filling volume

bottle volume

250 mL

color

pale yellow

pH

3.7±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Alicyclobacillus spp.

General description

Alicyclobacillus is a food-spoiling microorganism and requires an acidic environment to grow. YSG media supports the growth of all currently known species of Alicyclobacillus (A. acidocaldarius, A. acidoterrestris, A. cycloheptanicus, and A. hesperidium). The low pH value of the media, in combination with the high incubation temperature, inhibits the growth of contaminating microbiota. K Agar, when incubated at 45ºC, predominantly supports the growth of A. acidoterrestris and limits the growth of other species of the same genus. Therefore, K Agar is mainly used to detect A. acidoterrestris strains.

Application

M-YSG Broth is used for the detection and enrichment of Alicyclobacillus, in fruit juices and other acidic foods.

Components

  • 1 box with 10 bottles (250 mL)
  • Injectable cap: Plastic screw inner cap

Other Notes

Using syringe needles with a diameter greater than 0.8 mm is not recommended.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service