Skip to Content
Merck
All Photos(1)

Key Documents

C9595

Millipore

Chapman Stone Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

form

powder

Quality Level

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

application(s)

bioburden testing
environmental
food and beverages

microbiology

suitability

Staphylococcus spp.

Application

Recommended for the selective isolation of Staphylococci causing food poisoning.

Components

Ingredients (g/L):
Casein enzymic hydrolysate, 10.00
Yeast extract, 2.50
Gelatin, 30.00
D-Mannitol, 10.00
Sodium chloride, 55.00
Ammonium sulphate, 75.00
Dipotassium phosphate, 5.00
Agar, 15.00

Preparation Note

Suspend 20.25 grams of Chapman Stone Agar in100 mls of distilled water. Boil to dissolve the mediumcompletely. Sterilize by autoclaving at 15 lbs. pressure(121 °C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service