Skip to Content
Merck
All Photos(1)

Key Documents

B0382

Supelco

Bamethane hemisulfate salt

analytical standard

Synonym(s):

α-(Butylamino)methyl-p-hydroxybenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(OH)CH2NH(CH2)3CH3 · 1/2 H2SO4
CAS Number:
Molecular Weight:
258.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

OS(O)(=O)=O.CCCCNCC(O)c1ccc(O)cc1.CCCCNCC(O)c2ccc(O)cc2

InChI

1S/2C12H19NO2.H2O4S/c2*1-2-3-8-13-9-12(15)10-4-6-11(14)7-5-10;1-5(2,3)4/h2*4-7,12-15H,2-3,8-9H2,1H3;(H2,1,2,3,4)

InChI key

PARMADWNFXEEFC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service