Skip to Content
Merck
All Photos(1)

Key Documents

93967

Sigma-Aldrich

Peanut oil hardened

tested according to Ph. Eur.

Synonym(s):

Arachidis oleum hydrogenatus, Arachidis oleum hydrogenatum, Arachis oil hardened

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

paste

application(s)

pharmaceutical (small molecule)

lipid type

oils

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C30H45N9O5/c31-15-5-4-9-23(26(33)41)37-29(44)25(18-19-7-2-1-3-8-19)39-28(43)24(10-6-16-36-30(34)35)38-27(42)22(32)17-20-11-13-21(40)14-12-20/h1-3,7-8,11-14,22-25,40H,4-6,9-10,15-18,31-32H2,(H2,33,41)(H,37,44)(H,38,42)(H,39,43)(H4,34,35,36)/t22-,23-,24+,25-/m0/s1

InChI key

UEVAHGMTRWGMTB-JBXUNAHCSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service