Skip to Content
Merck
All Photos(1)

Key Documents

92760

Sigma-Aldrich

(Trimethylsilyl)isocyanate

technical, ≥95% (GC)

Synonym(s):

Isocyanatotrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiNCO
CAS Number:
Molecular Weight:
115.21
Beilstein:
1744962
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.396 (lit.)
n20/D 1.397

bp

90-92 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O
C[Si](C)(C)N=C=O

InChI

1S/C4H9NOSi/c1-7(2,3)5-4-6/h1-3H3

InChI key

NIZHERJWXFHGGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may become turbid on storage

Other Notes

Versatile reactive organic isocyanate; preparation of primary amines from Grignard reagents or from organo-lithiums[1][2]

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service