Skip to Content
Merck
All Photos(1)

Documents

91213

Supelco

Neochlorogenic acid

analytical standard

Synonym(s):

5-O-(trans-3,4-Dihydroxycinnamoyl)-D-quinic acid, trans-5-O-Caffeoylquinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1C[C@@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@H]1O)C(O)=O

InChI

1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14+,16-/m1/s1

InChI key

CWVRJTMFETXNAD-NXLLHMKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neochlorogenic acid is a phenolic acid, which is commonly found in apricots. It belongs to the category of hydroxycinnamates and may possess antioxidant property.It is reported to inhibit the oxidation of low-density lipoprotein, thereby reducing the risk of heart diseases.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Skinner M and Hunter D et al.
Bioactives in Fruit: Health Benefits and Functional Foods (2013)
Phenolic composition and antioxidant activity of prunes and prune juice (Prunus domestica)
Donovan.LJ, et al. et al.
Journal of Agricultural and Food Chemistry, 46(4), 1247-1252 (1998)
Liangmian Chen et al.
Chemical & pharmaceutical bulletin, 63(1), 25-32 (2015-03-07)
For the determination of seven caffeoylquinic acids [neochlorogenic acid (NcA), cryptochlorogenic acid (CcA), chlorogenic acid (CA), caffeic acid (CfA), isochlorogenic acid A (Ic A), isochlorogenic acid B (Ic B), isochlorogenic acid C (Ic C)] and two flavonoids [luteolin 7-O-glucoside (LtG) and luteolin (Lt)]
Liangmian Chen et al.
Journal of pharmaceutical and biomedical analysis, 95, 93-101 (2014-03-25)
A novel three-channel isocratic elution high performance liquid chromatography with electrochemical detection (3LC-ECD) system was developed for the simultaneous determination of various bioactive redox components in Shuang-Huang-Lian (SHL) preparations. This 3LC-ECD system consists of three isocratic elution flow ways, two
Fatih Mehmet Yılmaz et al.
Journal of food science and technology, 52(5), 2851-2859 (2015-04-22)
There is an increasing interest to bio-components obtained from fruit & vegetable wastes. Anthocyanin is one of the components regained from fruits especially from red ones. Sour cherry is considered as industrial fruit since most of the grown fruit is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service