Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 g | Please contact Customer Service for Availability | ₪1,651.00 |
About This Item
sterility
non-sterile
Quality Segment
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 13.5 g/L , casein enzymic hydrolysate, 4.0 g/L , chromogenic mixture, 2.5 g/L , peptic digest of animal tissue, 15.0 g/L
manufacturer/tradename
NutriSelect® Plus
final pH
7.2±0.2 (25 °C)
application(s)
bioburden testing
clinical testing
environmental
food and beverages, microbiology
storage temp.
2-8°C
suitability
Enterococcus spp., Escherichia coli, Klebsiella spp., Proteus spp., Pseudomonas spp., Salmonella spp., Staphylococcus spp., coliforms
Application
Preparation Note
Other Notes
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form powder | form powder | form powder |
| suitability Enterococcus spp., Klebsiella spp., Pseudomonas spp., Staphylococcus spp., Escherichia coli, Salmonella spp., Proteus spp., coliforms | suitability selective and differential for Salmonella spp., selective and differential for enterobacteriaceae | suitability Lactococcus spp., Leuconostoc spp. | suitability enterobacteriaceae, nonselective and differential for Escherichia coli, nonselective and differential for Klebsiella spp., nonselective and differential for Proteus spp., nonselective and differential for Pseudomonas spp., nonselective and differential for Staphylococcus spp., nonselective and differential for coliforms, nonselective and differential for enterobacteriaceae |
| composition agar, 13.5 g/L , chromogenic mixture, 2.5 g/L , casein enzymic hydrolysate, 4.0 g/L , peptic digest of animal tissue, 15.0 g/L | composition agar, 12.0 g/L , casein enzymic hydrolysate, 8.0 g/L , chromogenic mixture, 4.32 g/L , lactose, 3.0 g/L , neutral red, 0.02 g/L , peptic digest of animal tissure, 4.0 g/L , sodium chloride, 5.0 g/L , sodium deoxycholate, 1.0 g/L , yeast extract, 5.0 g/L | composition agar, 15.0 g/L , calcium lactate pentahydrate, 8.0 g/L , carboxymethyl cellulose (CMC), 0.40 g/L , casein enzymic hdrolysate, 18.0 g/L , chromogenic substrate (X-gal), 0.20 g/L , gelatine, 2.25 g/L , glucose, 4.5 g/L , lactose, 4.5 g/L , sodium chloride, 3.6 g/L , tricalcium dicitrate tetrahydrate, 6.65 g/L , trisodium citrate dihydrate, 1.8 g/L , yeast extract, 4.5 g/L | composition agar, 15 g/L , beef extract, 4 g/L , casein enzymic hydrolysate, 4 g/L , chromogenic mixture, 12.44 g/L , peptic digest of animal tissue, 18 g/L |
| sterility non-sterile | sterility - | sterility non-sterile | sterility non-sterile |
| application(s) bioburden testing | application(s) clinical testing | application(s) bioburden testing | application(s) clinical testing |
| manufacturer/tradename NutriSelect® Plus | manufacturer/tradename NutriSelect® Plus | manufacturer/tradename NutriSelect® Plus | manufacturer/tradename NutriSelect® Plus |
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



