Skip to Content
Merck
All Photos(1)

Key Documents

81368

Supelco

Poly(vinyl alcohol)

analytical standard, for GPC, Mw 7,200

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard
for GPC

mol wt

Mn ~4200
Mp ~5600
Mw ~7200

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.71

format

neat

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fusako Kawai et al.
Archives of microbiology, 195(2), 131-140 (2012-12-25)
Scanning electron microscopy (SEM) shows remarkable morphological surface changes in Sphingopyxis sp. 113P3 cells grown in polyvinyl alcohol (PVA) but not in Luria-Bertani medium (LB) (Hu et al. in Arch Microbiol 188: 235-241, 2007). However, transmission electron microscopy showed no
Piotr Hanczyc et al.
The journal of physical chemistry. B, 117(10), 2947-2954 (2013-02-15)
A stretched poly(vinyl alcohol) (PVA) film provides a unique matrix that enables also short DNA oligonucleotide duplex to be oriented and studied by linear dichroism (LD). This matrix further allows controlling DNA secondary structure by proper hydration (A or B
Jie Xue et al.
Journal of photochemistry and photobiology. B, Biology, 119, 31-36 (2013-01-15)
Synthetic adhesives inspired by marine mussel have attracted great attention due to its excellent water-resistance and good biocompatibility. In this study, a photocrosslinkable bioadhesive containing 3,4-Dihydroxy-l-phenylalanine (DOPA) functional group, which is central to curing mussel adhesive proteins, was prepared by
Gang Wang et al.
PloS one, 6(11), e27605-e27605 (2011-11-24)
Mucosal vaccination has been demonstrated to be an effective means of eliciting protective immunity against aerosol infections of foot and mouth disease virus (FMDV) and various approaches have been used to improve mucosal response to this pathogen. In this study
Susnata Bera et al.
Journal of nanoscience and nanotechnology, 13(1), 300-305 (2013-05-08)
Polymer capped CdSe nanoparticles have been grown using a simple chemical process. Transmission electron microscopy study revealed the formation of isolated CdSe nanoparticles encapsulated in polyvinyl alcohol matrix. The optical band gap of nanoparticle has been found to be blue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service