789001BC
Gemini® 2-Lane Sample Rack Base
BioControl Systems, for use with Gemini®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
BioControl Systems
compatibility
for use with Gemini®
storage temp.
room temp
Legal Information
Gemini is a registered trademark of Stratec
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service