Skip to Content
Merck
All Photos(1)

Documents

54374

Supelco

Lithium acetoacetate

analytical standard

Synonym(s):

Acetoacetic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOLi
CAS Number:
Molecular Weight:
108.02
Beilstein:
5774841
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

[Li+].CC(=O)CC([O-])=O

InChI

1S/C4H6O3.Li/c1-3(5)2-4(6)7;/h2H2,1H3,(H,6,7);/q;+1/p-1

InChI key

UTLRZTUJSMCBHB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service