Skip to Content
Merck
All Photos(1)

Key Documents

53706

Supelco

Methyl butyrate

certified reference material, TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740743
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 30 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.5 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.385 (lit.)

bp

102-103 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCCC(=O)OC

InChI

1S/C5H10O2/c1-3-4-5(6)7-2/h3-4H2,1-2H3

InChI key

UUIQMZJEGPQKFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl butyrate is a clear colourless liquid which is slightly soluble in water. It adds flavour and odour to fruits and flowers. It has a characteristic pineapple odour.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

It was used as reference material in analysing FAME from different fat classes using proton transfer reaction mass spectrometry (PTR-MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Authentication of feeding fats: classification of animal fats, fish oils and recycled cooking oils.
Van Ruth, S. M., et al.
Anim. Feed Sci. Technol., 155.1, 65-73 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service