Skip to Content
Merck
All Photos(1)

Documents

47717

Sigma-Aldrich

Methyl 4-formylbenzoate

technical, ≥90% (HPLC)

Synonym(s):

Methyl benzaldehyde-4-carboxylate, Methyl terephthalaldehydate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC6H4CO2CH3
CAS Number:
Molecular Weight:
164.16
Beilstein:
775895
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

≥90% (HPLC)

bp

265 °C (lit.)

mp

59-63 °C (lit.)
59-63 °C

solubility

methanol: 0.1 g/mL, clear

SMILES string

[H]C(=O)c1ccc(cc1)C(=O)OC

InChI

1S/C9H8O3/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-6H,1H3

InChI key

FEIOASZZURHTHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service