Skip to Content
Merck
All Photos(1)

Documents

47170

Sigma-Aldrich

1-Fluoro-4-nitrobenzene

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4NO2
CAS Number:
Molecular Weight:
141.10
Beilstein:
606923
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

5.5 mmHg ( 70 °C)

grade

purum

Assay

≥98.0% (GC)

color

deep green-yellow

refractive index

n20/D 1.531 (lit.)

bp

205 °C (lit.)

mp

21 °C (lit.)

density

1.33 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)cc1

InChI

1S/C6H4FNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H

InChI key

WFQDTOYDVUWQMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Fluoro-4-nitrobenzene is a fluorinated building block. On reaction with H218O, it affords 4-nitro-1-[18O]phenol.

Application

1-Fluoro-4-nitrobenzene may be used in the preparation of the following:
  • 1-(2-[18F]fluoroethoxy)-4-nitrobenzene
  • 4-[4-[1,1-di[4-(4-aminophenoxy)phenyl]-ethyl]phenoxy]aniline and swelling-agent-modified montmorillonite/PAA (poly(amic acid)
  • 1-(4-nitrophenyl)- and 1-(2,4-dinitrophenyl)uracil derivatives
  • sulfonaphthoaminophenyl fluorescein (SNAPF), an imaging probe

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Guo et al.
The Biochemical journal, 294 ( Pt 3), 653-656 (1993-09-15)
Michaelis-Menten parameters for hydrolysis of seven aryl N-acetyl alpha-D-neuraminides by Vibrio cholerae neuraminidase at pH 5.0 correlate well with the leaving-group pKa (delta pK 3.0; beta 1g (V/K) = -0.73, r = -0.93; beta 1g (V) = -0.25; r =
Effect of reactivity of organics-modified montmorillonite on the thermal and mechanical properties of montmorillonite/polyimide nanocomposites.
Tyan H-L, et al.
Chemistry of Materials, 13(1), 222-226 (2001)
Joanna Shepherd et al.
Chemistry & biology, 14(11), 1221-1231 (2007-11-21)
The myeloperoxidase (MPO)-derived oxidant hypochlorous acid (HOCl/OCl(-)) is implicated in the pathogenesis of atherosclerosis and other inflammatory states. We have synthesized an imaging probe, sulfonaphthoaminophenyl fluorescein (SNAPF), that selectively reacts with HOCl. SNAPF detects HOCl produced by stimulated MPO-expressing cells
Jinhe Pan et al.
Nuclear medicine and biology, 40(6), 850-857 (2013-06-19)
2-[(18)F]Fluoroethoxy and 3-[(18)F]fluoropropoxy groups are common moieties in the structures of radiotracers used with positron emission tomography. The objectives of this study were (1) to develop an efficient one-step method for the preparation of 2-[(18)F]fluoroethanol (2-[(18)F]FEtOH) and 3-[(18)F]fluoropropanol (3-[(18)F]FPrOH); (2)
A convenient method for N-1 arylation of uracil derivatives.
Gondela A and Walczak K.
Tetrahedron Letters, 47(27), 4653-4657 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service