Skip to Content
Merck
All Photos(1)

Key Documents

40821

Supelco

Pinnatoxin G solution

20 μg/g (in 0.1% acetic acid in 50% acetonitrile, nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H63NO7
CAS Number:
Molecular Weight:
693.95
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g (in 0.1% acetic acid in 50% acetonitrile, nominal concentration)

format

single component solution

storage temp.

−20°C

SMILES string

C=C(C[C@H]1O[C@]2(O[C@@]3(O[C@@H]4CCC3)CC2)[C@](C)(O)CC1)CCCC5=NC[C@H](C)[C@@H](C)C[C@]65CCC(C=C)=C[C@]6([H])[C@@H]7O[C@]8(C4)C[C@@H](C)[C@@H](O)[C@H]7O8

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

For the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service