Skip to Content
Merck
All Photos(1)

Key Documents

37209

Supelco

Chromate test kit Quantofix®

3-100 mg/L (CrO42-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

Quality Level

specific analyte(s)

chromate

measuring range

3-100 mg/L (CrO42-)

application(s)

environmental

detection method

colorimetric

General description

Contents:
Kit including reagents and test sticks for 100 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 − 3 − 10 − 30 − 100 mg/L CrO42-

Other Notes

this product will be discontinued, please find the alternative product here :
110012

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service