Skip to Content
Merck
All Photos(2)

Key Documents

32981

Supelco

Sulfamethoxypyridazine-d3

analytical standard

Synonym(s):

4-Amino-N-(6-methoxy-d3-3-pyridazinyl)benzenesulfonamide, N1-(6-Methoxy-d3-3-pyridazinyl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D3H9N4O3S
CAS Number:
Molecular Weight:
283.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])Oc1ccc(NS(=O)(=O)c2ccc(N)cc2)nn1

InChI

1S/C11H12N4O3S/c1-18-11-7-6-10(13-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,15)/i1D3

InChI key

VLYWMPOKSSWJAL-FIBGUPNXSA-N

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Jesús Serrano et al.
Antibiotics (Basel, Switzerland), 9(4) (2020-04-16)
The introduction of antimicrobial residues in the food chain has a significant impact on human health. An innovative solution to avoid their presence in meat is the adaptation of current control methods for use with in vivo matrixes. Thus, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service