Skip to Content
Merck
All Photos(1)

Documents

32565

Supelco

1H-Benzotriazole-(ring-d4) solution

10 μg/mL in acetone, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4HN3
CAS Number:
Molecular Weight:
123.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+4

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c2[nH]nnc2c1[2H]

InChI

1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)/i1D,2D,3D,4D

InChI key

QRUDEWIWKLJBPS-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

1H-Benzotriazole-(ring-d4) has been used as an analytical standard for the determination of:
  • Polar benzotriazolic compounds in water samples by concurrent derivatization–dispersive liquid–liquid microextraction (DLLME) followed by gas chromatography–mass spectrometry (GC-MS).
  • Benzothiazoles in indoor samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of benzotriazoles in water samples by concurrent derivatization-dispersive liquid-liquid microextraction followed by gas chromatography-mass spectrometry.
Casado J, et al.
Journal of Chromatography A, 1336, 1-9 (2014)
Benzothiazoles in indoor air from Albany, New York, USA, and its implications for inhalation exposure.
Wan Y, et al.
Journal of Hazardous Materials, 311, 37-42 (2016)
Attilio Naccarato et al.
Journal of chromatography. A, 1338, 164-173 (2014-03-19)
This work proposes a new approach for the simultaneous determination of benzothiazoles, benzotriazoles and benzosulfonamides in different environmental matrices and human urine, using solid-phase microextraction coupled with gas-chromatography-triple quadrupole mass spectrometry (SPME-GC-QqQMS). The analytes object of this investigation have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service