Skip to Content
Merck
All Photos(1)

Key Documents

31267

Supelco

Trichloroacetic acid

PESTANAL®, analytical standard

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein:
970119
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

vapor density

<1 (vs air)

vapor pressure

1 mmHg ( 51 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.62 (lit.)

bp

196 °C (lit.)

mp

54-58 °C (lit.)

solubility

soluble 81.7 g/L at 20 °C (completely)

density

1.62 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hikmat Al-Ahmadie et al.
Cancer discovery, 4(9), 1014-1021 (2014-06-18)
Metastatic solid tumors are almost invariably fatal. Patients with disseminated small-cell cancers have a particularly unfavorable prognosis, with most succumbing to their disease within two years. Here, we report on the genetic and functional analysis of an outlier curative response
Alan Mortensen et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 18(1), 55-62 (2012-10-17)
The ratio of the nitric oxide synthase (NOS) cofactor tetrahydrobiopterin (BH(4)) to its oxidized form dihydrobiopterin (BH(2)) has been suggested as an index of endothelial dysfunction. Consequently, much effort has been put into preserving the in vivo equilibrium between these
Katharine Herrick-Davis et al.
Molecular pharmacology, 87(4), 660-673 (2015-01-23)
G protein-coupled receptors (GPCRs) are a prominent class of plasma membrane proteins that regulate physiologic responses to a wide variety of stimuli and therapeutic agents. Although GPCR oligomerization has been studied extensively in recombinant cells, it remains uncertain whether native
Ying Wang et al.
Diabetes, 63(8), 2643-2655 (2014-03-13)
After diabetes, the heart has a singular reliance on fatty acid (FA) for energy production, which is achieved by increased coronary lipoprotein lipase (LPL) that breaks down circulating triglycerides. Coronary LPL originates from cardiomyocytes, and to translocate to the vascular
Verena Pfahler et al.
The New phytologist, 197(1), 186-193 (2012-10-31)
The objective of this study was to investigate the isotopic composition of oxygen bound to phosphate (δ(18)O-PO(4)) in different phosphorus (P) pools in plant leaves. As a model plant we used soybean (Glycine max cv Toliman) grown in the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service