Skip to Content
Merck
All Photos(1)

Key Documents

30371

Supelco

Benzoate Ionophore I

Selectophore, function tested

Synonym(s):

N,N′,N″,N″′,N″″,N″″′-Hexakisdecyl[dibenzo[klm,z(aa)(ab)]-1,5,9,16,20,24-hexaazacyclotriacontane-1,5,9,16,20,24-hexayl]hexaacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C100H184N12O6
CAS Number:
Molecular Weight:
1650.61
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

SMILES string

CCCCCCCCCCNC(=O)CN1CCCN(CC(=O)NCCCCCCCCCC)Cc2ccc(CN(CCCN(CCCN(CC(=O)NCCCCCCCCCC)Cc3ccc(CN(CCC1)CC(=O)NCCCCCCCCCC)cc3)CC(=O)NCCCCCCCCCC)CC(=O)NCCCCCCCCCC)cc2

InChI

1S/C100H184N12O6/c1-7-13-19-25-31-37-43-49-67-101-95(113)85-107-73-55-77-109(87-97(115)103-69-51-45-39-33-27-21-15-9-3)81-91-59-63-93(64-60-91)83-111(89-99(117)105-71-53-47-41-35-29-23-17-11-5)79-57-75-108(86-96(114)102-68-50-44-38-32-26-20-14-8-2)76-58-80-112(90-100(118)106-72-54-48-42-36-30-24-18-12-6)84-94-65-61-92(62-66-94)82-110(78-56-74-107)88-98(116)104-70-52-46-40-34-28-22-16-10-4/h59-66H,7-58,67-90H2,1-6H3,(H,101,113)(H,102,114)(H,103,115)(H,104,116)(H,105,117)(H,106,118)

InChI key

DGWPXUIFCNZAMC-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric Response of Liquid Membrane Electrode Incorporated with Macrocyclic Polyamine towards Benzoate.
Bulgariu, L., et al.
Analytical Letters, 36, 1325-1334 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service