Skip to Content
Merck
All Photos(1)

Documents

23491

Sigma-Aldrich

4-Chlorobenzaldehyde

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CHO
CAS Number:
Molecular Weight:
140.57
Beilstein:
385858
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

bp

213-214 °C (lit.)

mp

45-50 °C (lit.)
45-50 °C

SMILES string

[H]C(=O)c1ccc(Cl)cc1

InChI

1S/C7H5ClO/c8-7-3-1-6(5-9)2-4-7/h1-5H

InChI key

AVPYQKSLYISFPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

oxidizes easily

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service