Skip to Content
Merck
All Photos(1)

Key Documents

08011

Sigma-Aldrich

(R)-2-Vinyloxirane

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(R)-3,4-Epoxy-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O
CAS Number:
Molecular Weight:
70.09
Beilstein:
5238465
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −21.0±1°, c = 1% in pentane(lit.)
[α]20/D −21.0±1°, c = 1% in pentane

refractive index

n20/D 1.417

bp

66 °C (lit.)

density

0.870 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C=C[C@@H]1CO1

InChI

1S/C4H6O/c1-2-4-3-5-4/h2,4H,1,3H2/t4-/m1/s1

InChI key

GXBYFVGCMPJVJX-SCSAIBSYSA-N

Other Notes

Used to prepare stereodiversified scaffolds in library synthesis[1]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-58.0 °F - closed cup

Flash Point(C)

-50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiffany Malinky Gierasch et al.
Organic letters, 5(5), 621-624 (2003-02-28)
The syntheses of stereodiverse libraries of 12 and 19 are reported, where each asterisk represents an independently varied stereocenter. These scaffolds provide additional templates for investigations of geometric diversity in library syntheses. Libraries of these N-Fmoc-amino acids were further functionalized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service