03597
MALDI validation set PS, PMMA, PDMS, PEG and PSS Mp ~4′300 - 6′500
About This Item
Recommended Products
shelf life
limited shelf life, expiry date on the label
Quality Level
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
storage temp.
2-8°C
General description
Components
This mixed kit contains one polystyrene, one polymethyl methacrylate, one polydimethylsiloxane, one polyethylene glycol and one polystyrene sulphonate, with Mp ranging from ~4,300 to ~6,500.
Each standard is characterized with GPC and MALDI-ToF-MS.
related product
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service