Skip to Content
Merck
All Photos(1)

Key Documents

21-1584

Sigma-Aldrich

Nitrate nitrogen standard solution

100 ppm NO3-

Synonym(s):

Potassium nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
form:
liquid

form

liquid

availability

available only in Japan

concentration

100 ppm NO3-

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Pradeep et al.
Australian dental journal, 57(4), 429-434 (2012-11-29)
Dentinal hypersensitivity has been defined as a short, sharp pain arising from exposed dentine as a result of various stimuli such as heat, cold, chemical, or osmotic, that cannot be ascribed to any other pathology. This study was conducted to
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
A Kakar et al.
The Journal of clinical dentistry, 23(2), 40-47 (2012-07-12)
This clinical study was designed to compare dentin hypersensitivity relief among subjects who brushed their teeth with a new toothpaste containing 8.0% arginine, calcium carbonate, and 1000 ppm fluoride as sodium monofluorophosphate (MFP) to subjects who brushed with a commercially
E-A Kouotou et al.
Annales de dermatologie et de venereologie, 139(10), 617-620 (2012-11-06)
A cutaneous adverse drug reaction is a skin eruption secondary to the intake of a drug, be it prescribed by a medical practitioner or taken as auto-medication for a given ailment. In this document we present an original case of
[Alterations in the structure and activity of microbial soil community upon mineral nitrogen supplementation].
I V Evdokimov et al.
Mikrobiologiia, 81(6), 803-809 (2013-04-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service