Skip to Content
Merck
All Photos(1)

Documents

YG-10ML-SAM

Sigma-Aldrich

YG, Anti-Toxo FFMU MS-415 10ML Sample

clone MS-415, from human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

human

Quality Level

antibody product type

primary antibodies

clone

MS-415, monoclonal

isotype

IgM

shipped in

wet ice

Specificity

Anti-p30 TOXO

Application

SAMPLE FOR EVALUATION ONLY

Caution

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1% (w/v) sodium azide.

Storage and Stability

2–8 °C

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service