Skip to Content
Merck
All Photos(1)

Key Documents

XX7104712

Millipore

Fluids Contamination Analysis Kit, 230 V/50 Hz

Conforms to standard contamination test procedures such as ARP-598 (hydraulic fluids), ASTM-D2276 (aviation turbine fuels) & federal standard 791a (lubricants, liquid fuels & related products)

Synonym(s):

FLUID CONTAM ANAL KIT 230V50HZ

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
₪20,089.00

₪20,089.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
₪20,089.00

About This Item

UNSPSC Code:
41116116
NACRES:
NB.24

₪20,089.00


Please contact Customer Service for Availability

AC/DC input

220 V, 50 Hz

sterility

non-sterile

application(s)

industrial qc

Application

Conforms to standard contamination test procedures such as ARP-598 (hydraulic fluids), ASTM-D2276 (aviation turbine fuels) & federal standard 791a (lubricants, liquid fuels & related products).

Components

  • Glass Filter Holder with stainless steel screen (47mm)
  • vacuum filtering flask (1L)
  • filter forceps
  • latex tubing (1/4in x 23in)
  • Solvent Filtering Dispenser
  • PeteriSlides
  • Chemical Duty Pump (230V/50Hz)
  • MF-Millipore membranes - 47mm, mixed cellulose esters, white/plain, white/gridded, black/gridded

Disclaimer

In the European Union, this product will be considered Waste Electric and Electronic Equipment (WEEE) at the end of its life.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service