Skip to Content
Merck
All Photos(1)

Documents

TANKLK002

Milli-Q

Water Sensor Extension

Water Sensor with Cable for high-flow water purification systems, To be connected in series to a primary water sensor (ZFWATDET4).

Synonym(s):

Water sensor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.14

material

POM body (machined)
stainless steel electrode

packaging

pkg of 1 unit

cable L

4 m

compatibility

for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The water sensor is an accessory for water purification systems. It enables shutdown of the feed water supply in case of an unlikely event of a leak. The sensor detects water spillage on the floor and automatically closes the stand-alone inlet solenoid valve to prevent a lab flood and to protect the surroundings from water damage.
The water sensor includes a 4-meter extension cable with an electrical connection, to be connected in series to a primary water sensor (ZFWATDET4). This allows installation of the water sensor extension 8 meters away from the system. A typical installation would have the primary water sensor on the bench and the sensor extension on the floor.

Application

Detects water spillage on the floor and automatically closes the stand-alone inlet solenoid valve to protect the surroundings from water damage

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service