Skip to Content
Merck
All Photos(1)

Documents

SYMBTBG01

Millipore

Silicone drain tubing 

Accessory for Steritest® Symbio pumps, 1.5 meters

Synonym(s):

Hose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

silicone

Agency

EP 2.6.1
JP 4.06
USP 71

manufacturer/tradename

Steritest®

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sample preparation
sterility testing

compatibility

for use with Steritest® Compact Pump
for use with Steritest® Equinox Isofit Pump
for use with Steritest® Equinox Pump
for use with Steritest® Integral Pump
standard for VHP

shipped in

ambient

General description

The silicone drain tube is an accessory for STERITEST® Symbio pumps. One end of the drain tubing is connected to the drain tray of the pump and the other end is directly placed into the waste container or isolator table drain (depending on the pump version). It is autoclavable (121°C for 30 minutes or 134°C for 10 minutes).

Application

The silicon drain tubing is an accessory for STERITEST® Symbio pumps and is used for liquid disposal.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service