Skip to Content
Merck
All Photos(1)

Key Documents

SX0445

Supelco

Sodium citrate, dihydrate

Synonym(s):

Citric Acid, tri-Sodium Salt, Dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302

Assay

99.0-101.0% (acidimetric)

Quality Level

form

crystals

impurities

≤0.0005% Heavy metals (as Pb)
≤0.0005% Iron (Fe)
≤0.001% Chloride (Cl)
≤0.003% Ammonium (NH4)
≤0.004% Sulfate (SO4)
≤0.005% Calcium (Ca)
≤0.005% Insoluble matter

pH

7.5-9.0 (25 °C, 50 g/L in H2O)

mp

150 °C (anhydrous substance)

solubility

720 g/L

storage temp.

room temp

Application

crystals GR ACS

Analysis Note

Ammonium (NH4):0.003% max
Assay (acidimetric):99.0-101.0%
Calcium (Ca):0.005% max
Chloride (Cl):0.001% max
Heavy metals (as Pb):0.0005% max
Insoluble matter:0.005% max
Iron (Fe):0.0005% max
Sulfate (SO4):0.004% max
pH (5%, water, 25°C):7.5-9.0

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jared A Mereness et al.
The American journal of pathology, 190(2), 426-441 (2019-12-16)
Collagen VI (COL6) is known for its role in a spectrum of congenital muscular dystrophies, which are often accompanied by respiratory dysfunction. However, little is known regarding the function of COL6 in the lung. We confirmed the presence of COL6

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service