Skip to Content
Merck
All Photos(1)

Documents

ST1505

Sigma-Aldrich

Anti-WIPI1 Mouse mAb (3C1)

liquid, clone 3C1, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

3C1, monoclonal

form

liquid

does not contain

preservative

species reactivity

rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WIPI1(55062)

General description

Purified mouse monoclonal antibody. Recognizes the ~45 kDa WIPI1 protein.
Recognizes the ~45 kDa WIPI1 protein in PC-12 cells.
This Anti-WIPI1 Mouse mAb (3C1) is validated for use in Immunoblotting for the detection of WIPI1.
WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a β-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI1, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids.

Immunogen

A recombinant polypeptide corresponding to amino acids 348-446 of human WIPI1, expressed as a GST fusion protein
NA

Application


Immunoblotting (1-5 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
PC-12 cells

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service