Skip to Content
Merck
All Photos(1)

Key Documents

PX1286B

Sigma-Aldrich

Polyethylene glycol

400

Synonym(s):

Poly(ethylene glycol), PEG 400, Carbowax, Polyethylene Glycol 400, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
₪434.00
19 L
₪4,546.00

₪434.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
₪434.00
19 L
₪4,546.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2

₪434.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

color

APHA: ≤18

pH

5-7 (20 °C, 100 g/L in H2O, neutral)

mp

5 °C

transition temp

flash point >200 °C

shipped in

ambient

storage temp.

room temp

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Flexible Hydrogelation for Tactile Sensing: Recent research highlights the application of polyethylene glycol in bioinspired flexible hydrogels with programmable properties, enhancing the capabilities for tactile sensing applications (Wang et al., 2024).
  • Ocular Drug Delivery: Polyethylene glycol has been utilized in a sterilizable platform based on crosslinked xanthan gum for the controlled release of polymeric micelles, aimed at delivering neuroprotective compounds to the posterior eye segment (Signorini et al., 2024).
  • Nanoscale Drug Delivery Systems: A study on the nanoscale evaluation of black phosphorus nanosheets functionalized with polyethylene glycol and drug-loaded gold nanoparticles emphasizes its potential in enhancing drug delivery system stability (Gunathilaka and Shimomura, 2024).
  • Amphiphilic Thermogels for Biomedical Use: Polyethylene glycol is a key component in the design of carbosilane dendritic networks for developing high-swelling amphiphilic thermogels, suitable for various biomedical applications (Muñoz-Sánchez et al., 2024).
  • Biomedical and Environmental Polymer Applications: The role of polyethylene glycol in synthesizing natural and synthetic polymers showcases its versatility in both biomedical and environmental applications, offering new opportunities for sustainable material development (Satchanska et al., 2024).

Analysis Note

ClarityTo pass testColor (APHA)18 maxFormLiquidInfrared SpectrumConforms to standardSulfated ash0.05% maxSuspended matterTo pass testWater solubilityTo pass testpH of a 5% solution at 25°C4.5-7.5

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service