Skip to Content
Merck
All Photos(1)

Key Documents

MAPK8960B

Millipore

MultiScreen Punch Kit for 4 mL vials

Radionucleotide counting by transfer of individual filters to tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

product line

MultiScreen®

manufacturer/tradename

MultiScreen®

shipped in

ambient

Components

includes multiple punch and 2 carrier racks for 4 mL vials

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service