MABX4180
Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 180
clone 180, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody product type
primary antibodies
clone
180, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG2a
UniProt accession no.
Gene Information
human ... IGFBP4(3487)
General description
Antibody recognizes the N-term of Human insulin like growth factor binding protein 4 (IGFBP-4). IGFBP-4 is a member of the Insulin like growth factor family and is responsible for binding insulin like growth factors I and II. Research has linked levels of C-term (CT-IGFBP-4) and N-term (NT-IGFBP-4) to certain cardiac events, offering potential as a novel cardiac biomarker
Application
Detect NT-IGFBP-4 using this mouse monoclonal Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 180, Cat. No. MABX4180-1KC, validated for use in Western Blotting and ELISA.
Physical form
Format: Purified
Other Notes
Concentration: Please refer to lot specific datasheet.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service