Skip to Content
Merck

Skip To

MABS485

Anti-Ido1, clone 8G11 Antibody

clone 8G11, from mouse

Synonym(s):

Ido1, Indoleamine 2,3-dioxygenase 1, IDO-1, Indoleamine-pyrrole 2,3-dioxygenase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
₪1,606.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
8G11, monoclonal
Application:
FACS, WB
Citations:
1

₪1,606.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8G11, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IDO1(3620)

General description

Indoleamine 2,3-dioxygenase (IDO) is an enzyme that is responsible for converting tryptophan to kynurenines. IDO is expressed by a wide variety of tissues and can be upregulated by interferon gamma. IDO modulates levels of the amino acid tryptophan, which is vital for cell growth, but is also involved in the suppression of the immune response. IDO may be involved in the suppression of the immune response to tumours and blocking the IDO pathway may be a potential target for immunotherapy.
~44 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Recombinant protein corresponding to mouse Ido1.

Application

Anti-Ido1, clone 8G11 Antibody is an antibody against Ido1 for use in Western Blotting, Flow Cytometry.
Western Blotting Analysis: A representative lot detected Ido1 in splenic dendric cells from WT or KO mouse tissue lysate (Oberdan Leo, Universite Libre de Bruxelles).
Flow Cytometry Analysis: A representative lot detected Ido1 in splenic dendric cells from mouse (Oberdan Leo, Universite Libre de Bruxelles).

Biochem/physiol Actions

This antibody does not cross react with Ido2.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in Ido1-Flag tag transfected HEK293T cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Ido1 in 10 µg of Ido1-Flag tag transfected HEK293T cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB10009MAB5412SAB2501926
Gene Information

human ... IDO1(3620)

Gene Information

human ... IDO1(3620)

Gene Information

human ... IDO1(3620)

Gene Information

human ... IDO1(3620)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

goat

species reactivity

mouse

species reactivity

human

species reactivity

human

species reactivity

human

clone

8G11, monoclonal

clone

1F8.2, monoclonal

clone

10.1, monoclonal

clone

polyclonal

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters