Skip to Content
Merck

Skip To

MABS1981

Anti-TfR2 Antibody, clone 9F8-1C11

clone 9F8 1C11, from mouse

Synonym(s):

Transferrin receptor protein 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
₪1,605.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
9F8 1C11, monoclonal
Application:
ICC, IP, WB
Citations:
-

₪1,605.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F8 1C11, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TFR2(7036)

General description

Transferrin receptor protein 2 (UniProt: Q9UP52; also known as TfR2) is encoded by the TFR2 gene (Gene ID: 70360 in human. TfR2 is a single-pass type II membrane glycoprotein that plays a critical role in iron metabolism. It has a large C-terminal ectodomain and a small N-terminal cytoplasmic domain and shares 45% amino acid sequence with extracellular region of transferrin receptor (TfR). This homodimeric membrane receptor binds 2 molecules of transferrin and is internalized into endosomes that are acidified, resulting in the release of iron from transferrin. TfR2 contains a cytoplasmic internalization motif similar to TfR, and has 2 cysteines, which form inter subunit disulfide bonds, in the ectodomain proximal to the transmembrane domain. TfR2 mediates cellular uptake of transferrin-bound iron in a non-iron dependent manner. Both TfR and TfR2 bind diferric transferrin better than apo-transferrin at neutral pH. However, TFR2 displays about 25-fold lower affinity for transferrin compared to TfR and its expression is not controlled by cellular levels of iron. TfR2 is also suggested to be involved in involved in hepatocyte function and erythrocyte differentiation. Mutations in TFR2 gene are known to cause hemochromatosis 3 that is characterized by an iron overload where excess iron is deposited in several organs leading to organ failure. (Ref.: Kawabata H., et al (1999). J. Biol. Chem. 274 (30): 20826-20832; Vogt, TM et al. (2003). Blood 101(5):2008-2014.
~108 kDa observed; 88.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: extracellular domain
Purified ectodomains of the human TfR2.

Application

Anti-TfR2, clone 9F8-1C11 Antibody, Cat. No. MABS1981, is a highly specific mouse monoclonal antibody, that targets Tfr2 and has been tested in Western Blotting, Immunoprecipitation, and Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected Tfr2 in Immunocytochmeistry applications (Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14; Zhao, N., et. al. (2013). Biochemistry. 52(19):3310-9).

Immunohistochemistry Analysis: A representative lot detected Tfr2 in Immunohistochemistry applications (Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14).

Western Blotting Analysis: A representative lot detected Tfr2 in Western Blotting applications (Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14; Byrne, S.L., et. al. (2013). PLoS One. 8(7):e70199; Zhao, N., et. al. (2013). Biochemistry. 52(19):3310-9).

Immunoprecipitation Analysis: A representative lot detected Tfr2 in Immunoprecipitation applications (Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14; Johnson, M.B., et. al. (2004). Blood. 104(13):4287-93).
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

Clone 9F8-1C11 sprcifically detects human TfR2 and does exhibit reactivity with TfR.

Physical form

Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Format: Purified

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Tfr2 in 10 µg of K562 cell lysaste.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2102414SAB4200398FCMAB207F
biological source

mouse

biological source

-

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

Gene Information

human ... TFR2(7036)

Gene Information

human ... TFR2(7036)

Gene Information

human ... TFRC(7037)
mouse ... Tfrc(22042)
rat ... Tfrc(64678)

Gene Information

human ... TFRC(7037)

clone

9F8 1C11, monoclonal

clone

polyclonal

clone

polyclonal

clone

Ber-T9, monoclonal

species reactivity

human

species reactivity

mouse, human, rabbit, rat

species reactivity

mouse, rat, human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MABS198104054839189142

Questions

Reviews

No rating value

Active Filters