Skip to Content
Merck
All Photos(2)

Documents

MABS1282

Sigma-Aldrich

Anti-phospho-GRASP65 (Thr220/Thr224) Antibody, clone LX108

clone LX108, from mouse

Synonym(s):

Golgi reassembly-stacking protein 1, Golgi peripheral membrane protein p65, Golgi reassembly-stacking protein of 65 kDa, GRASP65, pGRASP65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LX108, monoclonal

species reactivity

rat

should not react with

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pThr220/pThr224)

Gene Information

human ... GORASP1(64689)

General description

Golgi reassembly-stacking protein 1 (UniProt: O35254; also known as Golgi peripheral membrane protein p65, Golgi reassembly-stacking protein of 65 kDa, GRASP65) is encoded by the Gorasp1 (also known as Grasp65) gene in rat. GRASP65 is a Golgi structural protein and mediates cisternae stacking via self-aggregation as well as interactions with other Golgi membrane-docking proteins, such as GM130. Golgi fragmentation results from amyloid beta accumulation, which causes activation of Cdk5 and phosphorylation of GRASP65 resulting in Golgi fragmentation. At least eight GRASP65 phosphorylation sites have been identified. GRASP65 phosphorylation during mitosis and dephosphorylation after mitosis are required for Golgi disassembly and reassembly during the cell cycle. Golgi fragmentation in Alzheimer s disease brains accelerates amyloid precursor protein (APP) trafficking and further enhances amyloid peptide (Ab) production. Expression of non-phosphorylatable mutants of GRASP65 or the inhibition of Cdk5 is shown to rescue the Golgi structure and reduce Ab40 and Ab42 production. GRASP65 mutants are also reported to reduce Ab secretion by increasing alpha cleavage of the APP. (Ref.: Joshi G., et al. (2013). Proc. Natl. Acad. Sci. USA 111(13): E1230-E1239).

Specificity

Clone LX108 specifically recognizes GRASP65 phosphorylated on Thr220/Thr224.

Immunogen

His-tagged recombinant full-length wild type GRASP65 protein treated with mitotic cytosol.

Application

Detect GRASP65 using this mouse monoclonal Anti-phospho-GRASP65 (Thr220/Thr224) Antibody, clone LX108, Cat. No. MABS1282. It is used for Immunocytochemistry and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected phospho-GRASP65 (Thr220/Thr224) in Western Blotting applications (Tang, D., et. al. (2012). Biol Open. 1(12):1204-14; Joshi, G., et. al. (2013). Proc. Natl. Acad. Sci. USA. 111(13):E1230-9).

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho-GRASP65 (Thr220/Thr224) in HeLa cells stably expressing rat GRASP65-GFP.

Immunocytochemistry Analysis: A representative lot detected phospho-GRASP65 (Thr220/Thr224) in Immunocytochemistry applications (Tang, D., et. al. (2012). Biol Open. 1(12):1204-14).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-GRASP65 (Thr220/Thr224) in rat liver Golgi and mitotic Golgi fragments.

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .

Target description

~100 kDa observed. Endogenous band may run at 72 kDa and GFP-tagged will show a band at 100 kDa Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service