Skip to Content
Merck
All Photos(1)

Documents

MABN2313

Sigma-Aldrich

Anti-Frataxin Antibody, clone 4F9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

4F9
monoclonal

purified by

using protein G

species reactivity

human, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

isotype

IgG2bκ

UniProt accession no.

General description

Full-length human recombinant Frataxin.

Specificity

Clone 4F9 is a mouse monoclonal antibody that specifically detects mature and intermediate forms of human Frataxin.

Application

Western Blotting Analysis: A representative lot detected Frataxin in Western Blotting applications (Piquet, F., et. al. (2018). Mol Ther. 26(8):1940-1952; Li, L., et. al. (2018). Nucleic Acid Ther. 28(1):23-33).

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Frataxin in K562 cell lysate.

Physical properties

~14 and 18 kDa observed; 23.13 kDa calculated (full length). Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, pH7.4 with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service