Skip to Content
Merck
All Photos(1)

Key Documents

MABF29

Sigma-Aldrich

Anti-IL-16 Antibody, clone 14.1

clone 14.1, from mouse

Synonym(s):

interleukin 16 (lymphocyte chemoattractant factor), lymphocyte chemoattractant factor, pro-interleukin-16, neuronal interleukin 16, prointerleukin 16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
neutralization: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL16(3603)

General description

Originally known as lymphocyte chemoattractant factor (LCF), Interleukin-16 (IL-16) is a product of human peripheral mononuclear cells when antigen-stimulated or mitogen-stimulated. It is also produced by dendritic cells, epithelial cells, eosinophils, fibroblasts and mast cells. The pro-form of IL-16 is a 631 amino acid protein that is cleaved by caspase 3 in activated CD4+ T cells where it is then secreted and is accessible for binding CD4 receptors. IL-16 regulates and induces migratory response in CD4+ lymphocytes, eosinophils, and monocytes. It is a soluble ligand for CD4 and functions to prime responsiveness to IL-2 and IL-15 in CD4+ T-cells. Although Isoform 1’s function is thought to be as an anchor to membrane ion channels, Isoform 3 is more widely researched and characterized as component of T-cell cycle progression.

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to the C-terminal region of secreted human IL-16.

Application

Detect IL-16 using this Anti-IL-16 Antibody, clone 14.1 validated for use in WB, NEUT, IH, IC, FC.
Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in IH to detect IL-16 in naive kidney tissue (Wang, S., et al., 2008).

Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IC to detect IL-16 in spinal cord cells (Skundric, D., et al., 2005).

Flow Cytometry Analysis: A previous lot was used by an independent laboratory in FC to detect IL-16 in human PBMCs (McFadden, C., et al., 2007).

Neutralizing Activity Assay: 1–10 µg/mL of a previous lot was used by an independent laboratory at concentrations sufficient to neutralize bioactivity of IL-16 (Morgan, R., et al., 2007).
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blot in human serum.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected IL-16 in 10 µg of human serum.

Target description

~20 kDa observed.
Monomers of IL-16 migrate on SDS-PAGE in the range of 17–20 kDa (Zhang, Y., et al., 1998). Active IL-16 (clone 14.1) is detected at ~17 kDa in WB (Skundric, D., et al., 2005).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgGκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human serum

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Templin et al.
Oncotarget, 8(30), 49253-49263 (2017-05-18)
Multiple myeloma (MM) is an incurable hematologic malignancy emerging from a plasma cell clone located in the bone marrow and is characterized by a high rate of fatal relapses after initially effective treatment. We have previously identified Interleukin-16 (IL-16) as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service