Skip to Content
Merck
All Photos(3)

Key Documents

MABC713

Sigma-Aldrich

Anti-KEAP1, clone 7G4B10 Antibody

clone 7G4B10, from mouse

Synonym(s):

Kelch-like ECH-associated protein 1, Cytosolic inhibitor of Nrf2, INrf2, Kelch-like protein 19, KEAP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₪2,472.00

₪2,472.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
₪2,472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₪2,472.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7G4B10, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KEAP1(9817)

Related Categories

General description

Kelch-like ECH-associated protein 1, also known as Cytosolic inhibitor of Nrf2 or INrf2 or Kelch-like protein 19, and encoded by the gene name KEAP1 or INRF2 or KLHL19, acts as a substrate adapter protein for the E3 ubiquitin ligase complex and targets NFE2L2/NRF2 for ubiquitination and degradation by the proteasome. KEAP1 shuttles between the cytoplasm and the nucleus and is widely expressed with high levels found in skeletal muscle.

Immunogen

Purified recombinant fragment of human KEAP1 expressed in E. Coli

Application

Anti-KEAP1, clone 7G4B10 Antibody is an antibody against KEAP1 for use in IHC, flow cytometry & ELISA.
Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected KEAP1 in HeLa cells.

ELISA: A representative lot detected control Antigen (100 ng) and Antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected KEAP1 in bladder cancer and cerebellum tissues.

Target description

70 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 0.5% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service