Skip to Content
Merck
All Photos(1)

Documents

MAB8742

Sigma-Aldrich

Anti-Western Equine Encephalitis Antibody, clone 2A2C.3

ascites fluid, clone 2A2C.3, Chemicon®

Synonym(s):

WEE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2A2C.3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
inhibition assay: suitable (hemagglutination)

isotype

IgG2a

shipped in

dry ice

Specificity

McMillan strain. Cross-reactivity with WEE (Highlands J and Sindbis strains), EEE, Semliki Forest, VEE

Immunogen

McMillan strain

Application

Detect Western Equine Encephalitis using this Anti-Western Equine Encephalitis Antibody, clone 2A2C.3 validated for use in ELISA, IF, HI.
ELISA

Indirect Immunofluorescence

Hemagglutination Inhibition

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Liquid; no preservatives

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6/12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgit Hülseweh et al.
mAbs, 6(3), 718-727 (2014-02-13)
This study describes the development of the first neutralizing antibodies against Western equine encephalitis virus (WEEV), a member of the genus Alphavirus. WEEV is transmitted by mosquitoes and can spread to the human central nervous system, causing symptoms ranging from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service