Skip to Content
Merck
All Photos(1)

Documents

HTS134M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human GIP Glucagon Family Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123201
eCl@ss:
32161000

description

GPCR Class B
protein target GIP

Quality Level

technique(s)

radioligand binding assay (RLBA): suitable

UniProt accession no.

Gene Information

human ... GIPR(2696)

General description

FUNCTION: SwissProt: P48546 # This is a receptor for GIP. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase.
SIZE: 466 amino acids; 53157 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P48546 ## Belongs to the G-protein coupled receptor 2 family.

Application

Radioligand binding assay

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservRC:R[590]Catives. Packaging method: Membranes protein were adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Store at –80°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.

Analysis Note

Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I] GIP (Perkin Elmer#: NEX-402)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 8-fold signal:background with 125I-labeled GIP at 0.2 nM

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service