Skip to Content
Merck
All Photos(1)

Key Documents

FX0415

Supelco

Formaldehyde Solution

36%

Synonym(s):

Formaldehyde Solution, Formalin, Formaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

450 ML
₪528.00

₪528.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
450 ML
₪528.00

About This Item

Empirical Formula (Hill Notation):
CH2O
Molecular Weight:
30.03
UNSPSC Code:
12352501
application(s):
histology

₪528.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

concentration

≥36%

color

APHA: ≤10

transition temp

flash point 60 °C

application(s)

histology

shipped in

ambient

storage temp.

room temp

Application

36%
Fixative used in histopathology, either by itself or in various mixtures.

Analysis Note

Acidity: To pass test
Assay: 36% min
Color (APHA): 10 max
Description: To pass test
Form: Clear liquid
Identification: To pass test
Solubility: To pass test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service