Skip to Content
Merck

Skip To

EX0197

Ether-Methanol Solvent

GR

Synonym(s):

MTBE-MeOH solvent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
20 L
Please contact Customer Service for Availability
₪6,584.00

About This Item

UNSPSC Code:
41116107
Bp:
34.8 °C

₪6,584.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

concentration

2.25-2.75 % (v/v) (methanol GC)

impurities

≤0.05% Water (H2O)

evapn. residue

≤0.001%

color

APHA: ≤10

bp

34.8 °C

transition temp

flash point -45 °C

shipped in

ambient

storage temp.

room temp

General description

EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.

Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.

Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.

Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.

Application

GR

Analysis Note

Assay (methanol, GC)2.25-2.75% (v/v)Color (APHA)10 maxEvaporation residue0.001% maxFormClear liquidInfrared SpectrumConforms to standardWater (H2O)0.05% max

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
900516320390MX0475
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100, 300

form

liquid

form

liquid

form

liquid

form

liquid

impurities

≤0.05% Water (H2O)

impurities

-

impurities

H2SO4, passes test (darkened), MnO4- reducers, passes test, ≤0.0002 meq/g Titr. base, ≤0.0003 meq/g Titr. acid, ≤0.001% acetaldehyde, ≤0.001% acetone, ≤0.001% formaldehyde, ≤0.1% water

impurities

≤0.0002 meq/g Titrable base, ≤0.0003 meq/g Titrable acid, ≤0.001% Carbonyl (acetone,formaldehyde,acetaldehyde), ≤0.05% Water (H2O)

bp

34.8 °C

bp

64.7 °C (lit.)

bp

64.7 °C (lit.)

bp

64.7 °C (lit.)

evapn. residue

≤0.001%

evapn. residue

-

evapn. residue

≤0.001%

evapn. residue

≤2 ppm

shipped in

ambient

shipped in

-

shipped in

-

shipped in

ambient


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 2 - STOT SE 3

target_organs

Central nervous system, Eyes,Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-49.0 °F

flash_point_c

-45 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
EX0197-104061839420717

Questions

Reviews

No rating value

Active Filters