Skip to Content
Merck
All Photos(1)

Key Documents

DR1098

Sigma-Aldrich

Anti-ID1 Mouse mAb (1F7)

liquid, clone 1F7, Calbiochem®

Synonym(s):

Anti-Inhibitor of DNA Binding 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F7, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ID1(3397)

General description

Purified mouse monoclonal antibody. Recognizes the ~17 kDa ID1 protein.
Recognizes the ~17 kDa ID1 protein in Y79 and HeLa cells.
This Anti-ID1 Mouse mAb (1F7) is validated for use in ELISA, Immunoblotting, Immunocytochemistry for the detection of ID1.

Immunogen

Full-length, recombinant, human ID1 (aa 1-156), expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Positive Control
Y79 cells, HeLa cells

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service