material
mixed cellulose esters (MCE) membrane
polyester support
silicone O-ring
sterility
non-sterile
Sterilization Compatibility
autoclavable compatible
steam-in-place compatible
product line
Milligard®
feature
hydrophilic
technique(s)
prefiltration: suitable
L
30 in.
W
2.9 in.
filtration area
2.1 m2
matrix
Milligard®
pore size
0.5/0.2 μm nominal pore size
cartridge code
Code 7
Looking for similar products? Visit Product Comparison Guide
Legal Information
MILLIGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service