CD-125ML-RU
Anti-E from the clone MS-78, Research Use Only
Anti-E from the clone MS-78, Human IgG, For non-IVD/Research Use, pkg of 125 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
antibody form
unpurified
clone
MS-78, monoclonal
form
liquid
packaging
pkg of 125 mL
technique(s)
agglutination assay: suitable
isotype
IgG
shipped in
ambient
storage temp.
2-8°C
General description
Clarified cell culture supernatant from cell lines expressing Human IgG monoclonal blod typing antibodies. For non-IVD/ research use. Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Application
Bulk for non IVD/Research Use
For Non-IVD / Research use only
Disclaimer
Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%(w/v) sodium azide.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service