Skip to Content
Merck
All Photos(2)

Key Documents

ABS1515

Sigma-Aldrich

Anti-phospho NFkB p52, (Ser222) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear factor NF-kappa-B p100 subunit, DNA-binding factor KBF2, H2TF1, Lymphocyte translocation chromosome 10 protein, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, Oncogene Lyt-10, Lyt10, Nuclear factor NF-kappa-B p52 subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₪2,252.00

₪2,252.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
₪2,252.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₪2,252.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer222)

Gene Information

human ... NFKB2(4791)

General description

The p52 NF-kB subunit is induced through activation of the alternative (noncanonical) NF-kB pathway in response to a range of stimuli such as lymphotoxin-b, BAFF and CD40, as well as DNA damage. The p52 formation results from proteolytic processing of its precursor protein p100 as a consequence of IKKalpha-mediated phosphorylation. The p52 is most commonly found as a heterodimer with RelB, although other dimer combinations have been identified by ChIP analysis. Ser222 of NF-kappa B p52 is phosphorylated by the kinase GSK3beta. Phosphorylation at S222 appears to regulate not only p52 partner dimerization but also the p52 dependent activation or repression of Skp2 gene expression.

Immunogen

Epitope: RHD domain
KLH-conjugated linear peptide corresponding to the RHD domain of human phospho NFkB p52, (Ser222).

Application

Anti-phospho NFkB p52, (Ser222) is an antibody against phospho NFkB p52 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected phospho NFkB p52, (Ser222) in CD40L and NTL cell lysate (Neil D. Perkins, Newcastle University).

Quality

Evaluated by Western Blotting in cell lysate U2OS transfected with p52 WT.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho NFkB p52, (Ser222) in 10 µg of cell lysate U2OS transfected with p52 WT.

Target description

~52 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service