Skip to Content
Merck
All Photos(3)

Key Documents

ABF1169

Sigma-Aldrich

Anti-Pyrin

from rabbit, purified by affinity chromatography

Synonym(s):

Marenostrin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
₪864.00
100 μL
₪2,090.00

₪864.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
₪864.00
100 μL
₪2,090.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₪864.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MEFV(4210)

General description

Pyrin (UniProt: O15553; also known as Marenostrin) is encoded by the MEFV (also known as MEF, TRIM20) gene (Gene ID: 4210) in human. Pyrin is a homotrimeric protein that is involved in the regulation of innate immunity and the inflammatory response in response to IFNG/IFN-gamma. It organizes autophagic machinery by serving as a platform for the assembly of ULK1, Beclin 1/BECN1, ATG16L1, and ATG8 family members and recognizes specific autophagy targets, thus coordinating target recognition with assembly of the autophagic apparatus and initiation of autophagy. Pyrin acts as an autophagy receptor for the degradation of several inflammasome components, including CASP1, NLRP1 and NLRP3, hence it may prevent excessive IL-1B- and IL-18-mediated inflammation. Pyrin is expressed in peripheral blood leukocytes, particularly in mature granulocytes and to a lesser extent in monocytes, but not in lymphocytes. It is also detected in spleen, lung and muscle, probably as a result of leukocyte infiltration in these tissues. Its expression is also detected in several myeloid leukemic, colon cancer, and prostate cancer cell lines. Three isoforms of Pyrin have been described that are produced by alternative splicing. Isoform 1 associates with microtubules and with the filamentous actin of perinuclear filaments and peripheral lamellar ruffles. In pre-apoptotic cells, it co-localizes with PYCARD/ASC in large specks (inflammasome). Isoform 2 is shown to be localized to the nucleus.

Specificity

This rabbit polyclonal antibody detects Pyrin in human cells. It targets an epitope within 15 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human Pyrin, isoform 1.

Application

Anti-Pyrin, Cat. No. ABF1169, is a rabbit polyclonal antibody that detects human Pyrin and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Pyrin in human tonsil, human cerebral cortex, human skeletal muscle, and human pancreatic cancer tissues.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Pyrin in 10 µg of THP-1 cell lysate.

Target description

~82 kDa observed; 86.44 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brianna Cyr et al.
Methods in molecular biology (Clifton, N.J.), 2696, 223-238 (2023-08-14)
The inflammasome is a multiprotein complex that is responsible for mounting an innate immune response through the activation of caspase-1 and the cleavage of interleukin-1β. This multiprotein complex plays an important role in a variety of central nervous system (CNS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service