Skip to Content
Merck
All Photos(2)

Documents

ABE1409

Sigma-Aldrich

Anti-Lysine-Specific Demethylase 4C (JMJD2C)

from rabbit, purified by affinity chromatography

Synonym(s):

Gene amplified in squamous cell carcinoma 1 protein, GASC-1 protein, JmjC domain-containing histone demethylation protein 3C, Jumonji domain-containing protein 2C, Jmjd2C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... KDM4C(23081)

General description

Lysine-specific demethylase 4C (UniProt: Q9H3R0; also known as EC: 1.14.11.-; Gene amplified in squamous cell carcinoma 1 protein, GASC-1 protein, JmjC domain-containing histone demethylation protein 3C, Jumonji domain-containing protein 2C, Jmjd2c) is encoded by the KDM4C (also known as GASC1, JHDM3C, JMJD2C, KIAA0780) gene (Gene ID: 23081) in human. Jmjd2c is an ubiquitously expressed histone demethylase that specifically demethylates Lys-9 and Lys-36 residues of histone H3. It is shown to demethylate only the trimethylated H3 and does not affect mono- and dimethylated residues. It does neither demethylate histone Lys-4 or Lys-27 of histone H3 nor the Lys-20 of histone H4. Human Jmjd2C contains one jumonji N domain (aa 16-58), a catalytic jumonji C domain (aa 144-310), two PHD zinc finger regions (aa 689-865), and two TUDOR methylated-histone binding domains (aa 877-991). Jmjd2c, along with its closely demethylase Jmjd2b, plays a vital role in self-renewal of embryonic stem cells (ESC) and in generation of induced pluripotent stem cells. Full-length Jmjd2b and Jmjd2c are shown to restore the normal ESC growth phenotype, ESC colony morphology, and also SSEA-1 expression. Jmjd2c is reported to assist polycomb repressive complex (PRC) in transcriptional repression. It is overexpressed in several esophageal squamous cell carcinomas.

Specificity

This polyclonal antibody detects Lysine-specific demethylase 4C in multiple human and mouse cell types.

Immunogen

KLH-Conjugated linear peptide corresponding to 18 amino acids from the N-terminal half of human Lysine-specific demethylase 4C.

Application

Anti-Lysine-Specific Demethylase 4C (JMJD2C) Antibody, Cat. No. ABE1409, is a highly specific rabbit polyclonal antibody that targets Lysine-specific demethylase 4C and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 2 µg/mL from a representative lot detected Lysine-Specific Demethylase 4C (JMJD2C) in 10 µg of C2C12 and HEK293 nuclear extract.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Lysine-Specific Demethylase 4C (JMJD2C) in human tonsil, human cervical cancer, and human liver cancer tissues.

Quality

Evaluated by Western Blotting in BALB/3T3 clone A31 cell nuclear extract.

Western Blotting Analysis: 2 µg/mL of this antibody detected Lysine-Specific Demethylase 4C (JMJD2C) in 10 µg of BALB/3T3 clone A31 nuclear extract.

Target description

~120 kDa observed; 119.98 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service